Anti-EGR1 Rabbit Polyclonal Antibody
Catalog # ROCK600-401-693
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:early growth response 1
- Antigen symbol:EGR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Chimpanzee,Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:1958
- Antigen synonyms:TIS8|AT225|ZNF225|ZIF-268|NGFI-A|G0S30|KROX-24
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:This affinity-purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 94-108 (eqpyehltaesfpdi) of Human EGR-1.
- Purification:affinity-purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
This affinity purified antibody has been tested for use in ELISA, immunochemistry and western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band at ~58 kDa in size corresponding to EGR-1 by western blotting.
Recommended Dilutions: ELISA: 1:4000 - 1:16000; Western Blot: 1:500 - 1:3000; Immunochemistry: 2 µg/ml to 20 µg/ml;
Type: Primary
Antigen: EGR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Chimpanzee, Mouse