Anti-GRIN2B Rabbit Polyclonal Antibody
ROCK612-401-D93
- Antibody type:Primary
- Antigen name:Glutamate receptor, ionotropic, N-methyl D-aspartate 2B
- Antigen symbol:GRIN2B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid
- Gene ID:24410
- Antigen synonyms:GluN2B
- Modification:phospho-Ser1480
- Storage buffer:0,01 M HEPES, 0,15 M Sodium Chloride, pH 7,50,1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-NMDA R2B pS1480 Antibody was produced by repeated immunizations with a synthetic phospho-peptide corresponding to amino acid residues surrounding Ser1480.
- Purification:affinity-purified
- Size:100 µl
- Pk:100 µl
Anti-NMDA R2B pS1480 (Rabbit) antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 180 kDa in size corresponding to NMDAR NR2B subunit Protein phosphorylated at Ser1480.
Recommended Dilutions: Western Blot: 1:1000;
Type: Primary
Antigen: GRIN2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat