Order Entry
Italy
ContactUsLinkComponent
 
Anti-GRIN2B Rabbit Polyclonal Antibody
  ROCK612-401-C89
undefined
Anti-GRIN2B Rabbit Polyclonal Antibody
  ROCK612-401-C89

 

  • Antibody type:
    Primary
  • Antigen name:
    Glutamate receptor, ionotropic, N-methyl D-aspartate 2B
  • Antigen symbol:
    GRIN2B
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • ChIP:
    Yes
  • Gene ID:
    24410
  • Antigen synonyms:
    GluN2B
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Anti-NMDA NR2B Subunit pY1472 (Rabbit) antibody is suitable for use in Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 180 kDa in size corresponding to NMDAR NR2B subunit Protein phosphorylated at Tyr1472.

Recommended Dilutions: Western Blot: 1:1000;

Type: Primary
Antigen: GRIN2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat