Anti-JAG1 Rabbit Polyclonal Antibody
Catalog # ROCK200-401-698S
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:jagged 1
- Antigen symbol:JAG1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:182
- Antigen synonyms:AWS|AHD|JAGL1|HJ1|AGS|CD339
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:This Protein A purified antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to amino acids 110-125 of human Jagged-1Protein.
- Purification:Protein A purified
- Size:25 µl
- Pk:25 µl
Specifications
About this item
This Protein A purified antibody has been tested for use in ELISA, immunochemistry, immunofluorescence microscopy and western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 150 kDa in size.
Recommended Dilutions: ELISA: 1:20000 - 1:100000; Immunofluorescence: 1:200 - 1:1000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:100 - 1:500;
Type: Primary
Antigen: JAG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse