Order Entry
Italy
ContactUsLinkComponent
Anti-Keratin 19 Mouse Monoclonal Antibody [clone: A53-B/A2.26, Ks 19.1]
Anti-Keratin 19 Mouse Monoclonal Antibody [clone: A53-B/A2.26, Ks 19.1]
Catalog # USBI138560
Supplier:  US Biological
Anti-Keratin 19 Mouse Monoclonal Antibody [clone: A53-B/A2.26, Ks 19.1]
Catalog # USBI138560
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Keratin 19
  • Antigen symbol:
    KRT19
  • Clonality:
    Monoclonal
  • Clone:
    A53-B/A2.26, Ks 19.1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,05% BSA, 0,05% sodium azide.
  • Storage temperature:
    –20 °C
  • Concentration:
    ~0,2 mg/ml
  • Immunogen:
    Human breast cancer MCF-7 cells
  • Purification:
    purified by Protein G chromatography
  • Size:
    20 µg
  • Pk:
    20 µG

Specifications

About this item

Recommended dilution: Western Blot: 0.5-1 ?g/ml; ; Immunofluorescence: 1-2?g/ml; Immunoprecipitation: 1-2?g/500 ?g; Immunochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: Keratin 19
Clonality: Monoclonal
Clone: A53-B/A2.26, Ks 19.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human,