Order Entry
Italy
ContactUsLinkComponent
Anti-Calcitonin Rabbit Polyclonal Antibody
Anti-Calcitonin Rabbit Polyclonal Antibody
  USBIC0115-08H-1
Anti-Calcitonin Rabbit Polyclonal Antibody
  USBIC0115-08H-1

 

  • Antibody type:
    Primary
  • Antigen name:
    Calcitonin
  • Antigen symbol:
    CALCA
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, protebase, sodium azide.
  • Storage temperature:
    –20 °C
  • Immunogen:
    Calcitonin.
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    1 ml
  • Pk:
    1 mL

 

 

Recommended dilution: Immunochemistry: 1:200-1:1000

Type: Primary
Antigen: Calcitonin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: