Order Entry
Italy
ContactUsLinkComponent
Anti-Heat Shock Protein 70, P. falciparum Rabbit Polyclonal Antibody
Anti-Heat Shock Protein 70, P. falciparum Rabbit Polyclonal Antibody
Catalog # USBIH1830-95S-100
Supplier:  US Biological
Anti-Heat Shock Protein 70, P. falciparum Rabbit Polyclonal Antibody
Catalog # USBIH1830-95S-100
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Heat Shock Protein 70, P. falciparum
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,4, 0,09% sodium azide, 50% glycerol.
  • Storage temperature:
    –20 °C
  • Concentration:
    ~1 mg/ml
  • Immunogen:
    His-tagged and purified P. falciparum Hsp70 C-terminus.
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 μg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Immunofluorescence:1:50; Western Blot:1:2000

Type: Primary
Antigen: Heat Shock Protein 70, P. falciparum
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: