Anti-Endoplasmic Reticulum Protein 57 Mouse Monoclonal Antibody [clone: 10F699]
Nome del fornitore: US Biological
Certificati
Informazioni su questo articolo
Recommended dilution: Western Blot:1:2000
Type: Primary
Antigen: Endoplasmic Reticulum Protein 57
Clonality: Monoclonal
Clone: 10F699
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Dog, Guinea pig, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat,
Specifiche
- Catalog No:
- USBIE2291-75C-50
- USBIE2291-75C-200
- Antigen name:
- Endoplasmic Reticulum Protein 57
- Endoplasmic Reticulum Protein 57
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 10F699
- 10F699
- Reactivity:
- Hamster, Pig, Rabbit, Monkey, Human, Bovine, Guinea pig, Dog, Rat, Mouse
- Hamster, Pig, Rabbit, Monkey, Human, Bovine, Guinea pig, Dog, Rat, Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Immunogen:
- Human recombinant ERp57 (GRP58).
- Human recombinant ERp57 (GRP58).
- Form:
- liquid
- liquid
- Purification:
- purified by Protein G chromatography
- purified by Protein G chromatography
- Concentration:
- ~1 mg/ml
- ~1 mg/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, pH 7,4,
- PBS, pH 7,4,
- Storage temperature:
- –20 °C
- –20 °C
Specifiche
Frequently Bought Together