Order Entry
Italy
ContactUsLinkComponent
Anti-Substance P Mouse Monoclonal Antibody [clone: SP-DE4-21]
Anti-Substance P Mouse Monoclonal Antibody [clone: SP-DE4-21]
Catalog # USBIS8000-01B-10
Supplier:  US Biological
Anti-Substance P Mouse Monoclonal Antibody [clone: SP-DE4-21]
Catalog # USBIS8000-01B-10
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Substance P
  • Clonality:
    Monoclonal
  • Clone:
    SP-DE4-21
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Guinea pig,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    lyophilized powder
  • Storage buffer:
    PBS, pH 7,2, 0,1M glycine.
  • Storage temperature:
    –20 °C
  • Concentration:
    ~1 mg/ml (after reconstitution)
  • Immunogen:
    Synthetic peptide corresponding RPRPQQFFGLM from human Substance P.
  • Purification:
    purified by Protein A chromatography
  • Size:
    10 μg
  • Pk:
    10 µG

Specifications

About this item

Recommended dilution: ELISA: 1:10000 Immunochemistry: 1:300

Type: Primary
Antigen: Substance P
Clonality: Monoclonal
Clone: SP-DE4-21

Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Guinea pig, Human, Mouse, Rat,