Anti-Parathyroid Hormone Mouse Monoclonal Antibody [clone: A1/70]
Catalog # USBIP3109-10D-10
Nome del fornitore: US Biological
Specifiche
- Antibody type:Primary
- Antigen name:Parathyroid Hormone
- Clonality:Monoclonal
- Clone:A1/70
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:No
- Form:lyophilized powder
- Amino acid number:1 to 38
- Storage buffer:PBS, pH 7,2
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Synthetic human PTH (aa 1-38) polylysine conjugated (SVSEIQLMHNLGKHLNSMERVEWLRKKLQDVHNFVALG)
- Tested applications:Radio Immunoassay
- Purification:purified by Protein A chromatography
- Size:10 μg
- Pk:10 µG
Specifiche
Informazioni su questo articolo
Recommended dilution: Radio Immunoassay: 20ng/ml
Type: Primary
Antigen: Parathyroid Hormone
Clonality: Monoclonal
Clone: A1/70
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,