Order Entry
Italy
ContactUsLinkComponent
Anti-Glucagon-like Peptide 1 Mouse Monoclonal Antibody [clone: GLP-1/18]
Anti-Glucagon-like Peptide 1 Mouse Monoclonal Antibody [clone: GLP-1/18]
Supplier:  US Biological
Certificates
Anti-Glucagon-like Peptide 1 Mouse Monoclonal Antibody [clone: GLP-1/18]
Supplier:  US Biological

Specifications

  • Catalog No:
  • USBIG2040-35C-10
  • USBIG2040-35C-100
  • Antigen symbol:
  • GLP1
  • GLP1
  • Antigen name:
  • Glucagon-like peptide 1
  • Glucagon-like peptide 1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • GLP-1/18
  • GLP-1/18
  • Reactivity:
  • Pig, Human, Rat
  • Pig, Human, Rat
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Synthetic peptide corresponding to aa7-36 of human GLP-1 conjugated to KLH.
  • Synthetic peptide corresponding to aa7-36 of human GLP-1 conjugated to KLH.
  • Amino acid number:
  • 7 to 36
  • 7 to 36
  • Form:
  • lyophilized powder
  • lyophilized powder
  • Purification:
  • purified by Protein G chromatography
  • purified by Protein G chromatography
  • Concentration:
  • ~1 mg/ml (after reconstitution)
  • ~1 mg/ml (after reconstitution)
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • PBS, pH 7,4. 100ul sterile ddH2O.
  • PBS, pH 7,4. 100ul sterile ddH2O.
  • Storage temperature:
  • –20 °C
  • –20 °C
  • Tested applications:
  • Dot Blot, Electron Microscopy
  • Dot Blot, Electron Microscopy

Specifications

Related Information

Frequently Bought Together