Order Entry
Italy
ContactUsLinkComponent
Anti-Brain Natriuretic Peptide Mouse Monoclonal Antibody [clone: 21-6-6]
Anti-Brain Natriuretic Peptide Mouse Monoclonal Antibody [clone: 21-6-6]
Supplier:  US Biological
Certificates
Anti-Brain Natriuretic Peptide Mouse Monoclonal Antibody [clone: 21-6-6]
Supplier:  US Biological

Specifications

  • Catalog No:
  • USBIB2702-34A-10
  • USBIB2702-34A-100
  • Antigen symbol:
  • BNP
  • BNP
  • Antigen name:
  • Brain Natriuretic Peptide
  • Brain Natriuretic Peptide
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 21-6-6
  • 21-6-6
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG2a
  • IgG2a
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Synthetic human pro-BNP (aa 1-21) polylysine conjugated (HPLGSPGSASDLETSGLQEQR)
  • Synthetic human pro-BNP (aa 1-21) polylysine conjugated (HPLGSPGSASDLETSGLQEQR)
  • Amino acid number:
  • 1 to 21
  • 1 to 21
  • Sequence:
  • HPLGSPGSASDLETSGLQEQR
  • Form:
  • lyophilized powder
  • lyophilized powder
  • Purification:
  • purified by Protein A chromatography
  • purified by Protein A chromatography
  • Concentration:
  • ~1 mg/ml
  • ~1 mg/ml
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • 200mM Na Citrate-Tris, 500mM sodium chloride, pH 7,5, 0,02% sodium azide.
  • 200mM Na Citrate-Tris, 500mM sodium chloride, pH 7,5, 0,02% sodium azide
  • Storage temperature:
  • –20 °C
  • –20 °C

Specifications

Related Information

Frequently Bought Together