Anti-Atrial Natriuretic Peptide, pro- Mouse Monoclonal Antibody [clone: A138-M14-19-H3]
Supplier: US Biological
Certificates
About this item
Type: Primary
Antigen: Atrial Natriuretic Peptide, pro-
Clonality: Monoclonal
Clone: A138-M14-19-H3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Specifications
- Catalog No:
- USBIA4150-03J-100
- USBIA4150-03J-10
- Antigen name:
- Atrial Natriuretic Peptide, pro-
- Atrial Natriuretic Peptide, pro-
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- A138-M14-19-H3
- A138-M14-19-H3
- Host:
- Mouse
- Mouse
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic peptide corresponding to aa80-93, (C)SDRSALLKSKLRAL of human pro-ANP.
- Synthetic peptide corresponding to aa80-93, (C)SDRSALLKSKLRAL of human pro-ANP.
- Amino acid number:
- 80 to 93
- 80 to 93
- Sequence:
- CSDRSALLKSKLRAL
- Form:
- lyophilized powder
- lyophilized powder
- Purification:
- purified by Protein G chromatography
- purified by Protein G chromatography
- Concentration:
- ~1 mg/ml
- ~1 mg/ml
- Cross adsorption:
- No
- No
- Storage temperature:
- –20 °C
- –20 °C
Specifications
Frequently Bought Together