Anti-Atrial Natriuretic Peptide, pro- Mouse Monoclonal Antibody [clone: A137-M14-19-F9]
Type: Primary
Antigen: Atrial Natriuretic Peptide, pro-
Clonality: Monoclonal
Clone: A137-M14-19-F9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
- Catalog No:
- USBIA4150-03F-100
- USBIA4150-03F-10
- Antigen name:
- Atrial Natriuretic Peptide, pro-
- Atrial Natriuretic Peptide, pro-
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- A137-M14-19-F9
- A137-M14-19-F9
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic peptide corresponding to aa40-55, (C)EPNEEAGAALSPLPEV of human pro-ANP.
- Synthetic peptide corresponding to aa40-55, (C)EPNEEAGAALSPLPEV of human pro-ANP.
- Amino acid number:
- 40 to 55
- 40 to 55
- Sequence:
- CEPNEEAGAALSPLPEV
- Form:
- lyophilized powder
- lyophilized powder
- Concentration:
- ~1 mg/ml
- ~1 mg/ml
- Cross adsorption:
- No
- No
- Storage temperature:
- –20 °C
- –20 °C
Frequently Bought Together