Order Entry
Italy
ContactUsLinkComponent
Anti-Atrial Natriuretic Peptide, pro- Mouse Monoclonal Antibody [clone: A136-10-L13-10-H8]
Anti-Atrial Natriuretic Peptide, pro- Mouse Monoclonal Antibody [clone: A136-10-L13-10-H8]
Catalog # USBIA4150-03C-10
Supplier:  US Biological
Anti-Atrial Natriuretic Peptide, pro- Mouse Monoclonal Antibody [clone: A136-10-L13-10-H8]
Catalog # USBIA4150-03C-10
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Atrial Natriuretic Peptide, pro-
  • Clonality:
    Monoclonal
  • Clone:
    A136-10-L13-10-H8
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    lyophilized powder
  • Amino acid number:
    12 to 25
  • Storage temperature:
    –20 °C
  • Concentration:
    ~1 mg/ml
  • Immunogen:
    Synthetic peptide corresponding to aa12-25 (LMDFKNLLDHLEEK(C) of human pro-ANP.
  • Purification:
    purified by Protein G chromatography
  • Size:
    10 μg
  • Pk:
    10 µG

Specifications

About this item

Type: Primary
Antigen: Atrial Natriuretic Peptide, pro-
Clonality: Monoclonal
Clone: A136-10-L13-10-H8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,