Anti-CREB (Ser133), Mouse Monoclonal Antibody [clone: 6D323]
USBIC7915-04B
- Antibody type:Primary
- Antigen symbol:CREB
- Clonality:Monoclonal
- Clone:6D323
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Amino acid number:125 to 135
- Modification:phospho-Ser133
- Immunogen:Synthetic peptide corresponding to aa 125-135 of CREB phosphorylated at Ser133.
- Size:50 µg
- Pk:100 µG
Type: Primary
Antigen: CREB (Ser133),
Clonality: Monoclonal
Clone: 6D323
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Mouse, Rat,