Anti-Interleukin 19 Rabbit Polyclonal Antibody (Biotin)
Supplier: US Biological
Certificates
About this item
Recommended dilution: ELISA: 0.15-0. 30 ?g/ml
Type: Primary
Antigen: Interleukin 19
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,
Specifications
- Catalog No:
- USBII8441-03A-25
- USBII8441-03A-50
- Antigen symbol:
- IL19
- IL19
- Antigen name:
- Interleukin 19
- Interleukin 19
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Highly pure (> 98%) recombinant human Interleukin-19
- Highly pure (> 98%) recombinant human Interleukin-19
- Form:
- lyophilized powder
- lyophilized powder
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Concentration:
- ~0,05 mg/ml
- ~0,05 mg/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS
- PBS
- Storage temperature:
- –20 °C
- –20 °C
Specifications
Frequently Bought Together