Anti-Fibroblast Growth Factor 17 Goat Polyclonal Antibody
Catalog # USBIF4212-76C-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Fibroblast growth factor 17
- Antigen symbol:FGF17
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:lyophilized powder
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Highly pure (> 98%) recombinant human FGF-17
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ELISA: >0.5 ?g/ml
Type: Primary
Antigen: Fibroblast Growth Factor 17
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human,