Order Entry
Italy
ContactUsLinkComponent
Anti-Connexin 40 Rabbit Polyclonal Antibody
Anti-Connexin 40 Rabbit Polyclonal Antibody
Catalog # USBIC7856-02F
Supplier:  US Biological
Anti-Connexin 40 Rabbit Polyclonal Antibody
Catalog # USBIC7856-02F
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Connexin 40
  • Antigen symbol:
    Cx40
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    PBS, pH 7,2, 0,01% BSA, 0,05% sodium azide,before the addition of glycerol to 40%.
  • Storage temperature:
    –20 °C
  • Concentration:
    ~1 mg/ml
  • Immunogen:
    Synthetic peptide of C-terminus, cytoplsmic domain of rat Connexin 40. Peptide. (KLH).
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Recommended dilution: Western Blot: 1-10 ?g/ml

Type: Primary
Antigen: Connexin 40
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat,