Anti-Ki-67 Rabbit Polyclonal Antibody
USBIK1700-05C-100
- Antibody type:Primary
- Antigen symbol:KI67
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Ki-67
- Storage buffer:10mM PBS, pH 7.6, 0,2% BSA and 15mM sodium azide before the addition of glycerol to 40%.
- Storage temperature:–20 °C
- Concentration:~0,1 mg/ml
- Immunogen:A synthetic peptide from C-terminus of human Ki67. Cellular Localization: Nuclear.
- Purification:purified by immunoaffinity chromatography
- Size:100 μl
- Pk:100 µl
Recommended dilution: Immunochemistry: 1:150
Type: Primary
Antigen: Ki-67
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat,