Anti-SVR4-like Rabbit Polyclonal Antibody
AGRIAS132726
: Agrisera
- Antibody type:Primary
- Antigen symbol:SVR4-like
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:A. thaliana,H. vulgare
- Western blot:Yes
- Cross adsorption:No
- UniProtKB:Q84RF5
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:KLH-conjugated synthetic peptide derived from Arabidopsis thaliana SVR4-like protein sequence,
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: SVR4-like
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: A. thaliana, H. vulgare