Anti-IgG (H+L) Goat Polyclonal Antibody
Catalog # AGRIAS10761
Supplier: Agrisera
Specifications
- Antibody type:Secondary
- Antigen symbol:IgG (H+L)
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Reactivity:Rabbit,Human,Bovine,Mouse
- Cross adsorption:No
- Storage temperature:+2°C to +8 °C
- Shipping temperature:Ambient
- Immunogen:purified human IgG (H&L)
- Purification:affinity-purified
- Size:2 mg
- Pk:2 mg
Specifications
About this item
Type: Secondary
Antigen: IgG (H+L)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Bovine, Human, Mouse, Rabbit