Anti-Hcn1 Mouse Monoclonal Antibody [clone: S70-28]
Catalog # ORIGTA326543
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:HCN1
- Clonality:Monoclonal
- Clone:S70-28
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:No
- Form:Liquid
- Amino acid number:778 to 910
- Storage buffer:PBS pH 7,4, 50% glycerol
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Fusion protein amino acids 778-910 (C terminus) of rat HCN1,
- Purification:purified by Protein G chromatography
- Size:100µg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1-10?g/ml; Immunochemistry: 0.1-1.0?g/ml; Immunofluorescence: 1.0-10?g/ml
Type: Primary
Antigen: Hcn1
Clonality: Monoclonal
Clone: S70-28
Conjugation: Unconjugated
Epitope: C-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat