Anti-ATP7B Mouse Monoclonal Antibody [clone: S62-29]
Catalog # ORIGTA326438
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:ATPase, Cu++ transporting, beta polypeptide
- Antigen symbol:ATP7B
- Clonality:Monoclonal
- Clone:S62-29
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:Liquid
- Amino acid number:3 to 21
- Storage buffer:PBS pH 7,4, 50% glycerol, 0,09% sodium azide
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Synthetic peptide amino acids 3-21 (cytoplasmic N-terminus) of human Copper-transporting ATPase2
- Purification:purified by Protein G chromatography
- Size:100µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: ATP7B
Clonality: Monoclonal
Clone: S62-29
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat