Order Entry
Italy
ContactUsLinkComponent
Anti-SLC1A1 Mouse Monoclonal Antibody [clone: S180-41]
Anti-SLC1A1 Mouse Monoclonal Antibody [clone: S180-41]
Catalog # ORIGTA326381
Nome del fornitore:  OriGene
Anti-SLC1A1 Mouse Monoclonal Antibody [clone: S180-41]
Catalog # ORIGTA326381
Nome del fornitore:  OriGene

Specifiche

  • Antibody type:
    Primary
  • Antigen symbol:
    Solute carrier family 1, member 1
  • Clonality:
    Monoclonal
  • Clone:
    S180-41
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Epitope:
    Full length
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    PBS pH 7,4, 50% glycerol
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Fusion protein amino acids 1-524 (full length) of human EAAC1
  • Purification:
    purified by Protein G chromatography
  • Size:
    100µg
  • Pk:
    100 µG

Specifiche

Informazioni su questo articolo

Type: Primary
Antigen: SLC1A1
Clonality: Monoclonal
Clone: S180-41
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human