Anti-UCHL1 Rabbit Polyclonal Antibody
Catalog # ORIGTA324710
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Ubiquitin Carboxyl-Terminal Esterase L1
- Antigen symbol:UCHL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Pig,Monkey,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Epitope:near the C-terminus
- Cross adsorption:No
- Form:Liquid
- Amino acid number:187 to 214
- Storage buffer:Purified polyclonal antibody supplied in PBS with 0,09% (W/V) sodium azide.
- Molecular weight:24.82 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:This UCHL1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 187-214 amino acids from the C-terminal region of human UCHL1.
- Purification:purified by Protein chromatography, followed by peptide affinity purification
- Size:400 μl
- Pk:400 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:10-50; Immunochemistry: 1:10-50
Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Bovine, Human, Monkey, Mouse, Pig, Rat