Anti-PTP4A2 Rabbit Polyclonal Antibody
Catalog # ORIGTA324079
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Protein tyrosine phosphatase type IVA, member 2
- Antigen symbol:PTP4A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Amino acid number:corresponding to a region between amino acids 2 to 164
- Storage buffer:PBS pH 7,3, 0,05% NaN3, 50% glycerol
- Molecular weight:19 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Fusion protein corresponding to a region derived from 2-164 amino acids of human protein tyrosine phosphatase type IVA, member 2
- Purification:purified by antigen affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: ELISA: 1:2000-5000; Western Blot: 1:200-1000; Immunochemistry: 1:25-100
Type: Primary
Antigen: PTP4A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat