Anti-ACOX1 Rabbit Polyclonal Antibody
Catalog # ORIGTA324048
Nome del fornitore: OriGene
Specifiche
- Antibody type:Primary
- Antigen name:acyl-CoA oxidase 1, palmitoyl
- Antigen symbol:ACOX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:Liquid
- Antigen synonyms:AOX
- Storage buffer:PBS pH 7,3, 0,05% NaN3, 50% glycerol
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Fusion protein corresponding to C terminal 250 amino acids of human acyl-CoA oxidase 1, palmitoyl
- Purification:purified by antigen affinity chromatography
- Size:100 μl
- Pk:100 µl
Specifiche
Informazioni su questo articolo
Recommended Dilutions: ELISA: 1:2000-5000; Immunochemistry: 1:50-200
Type: Primary
Antigen: ACOX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat