Anti-SPN Mouse Polyclonal Antibody [clone: eBio84-3C1 (84-3C1)]
ORIGTA320465
: OriGene
- Antibody type:Primary
- Antigen symbol:SPN
- Clonality:Polyclonal
- Clone:eBio84-3C1 (84-3C1)
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:Aqueous buffer, 0,09% sodium azide, may contain carrier protein/stabilizer
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Type: Primary
Antigen: SPN
Clonality: Polyclonal
Clone: eBio84-3C1 (84-3C1)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human