Order Entry
Italy
ContactUsLinkComponent
Anti-LAT Mouse Polyclonal Antibody [clone: LAT.10-17 (10-17)]
Anti-LAT Mouse Polyclonal Antibody [clone: LAT.10-17 (10-17)]
Catalog # ORIGTA320411
Supplier:  OriGene
Anti-LAT Mouse Polyclonal Antibody [clone: LAT.10-17 (10-17)]
Catalog # ORIGTA320411
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    LAT
  • Clonality:
    Polyclonal
  • Clone:
    LAT.10-17 (10-17)
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Storage buffer:
    Aqueous buffer, 0,09% sodium azide, may contain carrier protein/stabilizer
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Purification:
    purified by affinity chromatography
  • Size:
    25 μg
  • Pk:
    25 µG

Specifications

About this item

Type: Primary
Antigen: LAT
Clonality: Polyclonal
Clone: LAT.10-17 (10-17)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human