Anti-DsRed-Express2 IgG2b Monoclonal Antibody (Magnetic beads) [clone: OTI1C4 (formerly 1C4)]
ORIGTA183029
: OriGene
- Antibody type:Tag
- Antigen symbol:DsRed-Express2
- Clonality:Monoclonal
- Clone:OTI1C4 (formerly 1C4)
- Conjugation:Magnetic beads
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Form:Concentrated
- Storage buffer:PBS, pH 7,4, containing 0,1% BSA, 0,02% NaN3 and 50% glycerol with final concentration 5mg/mL beads
- Molecular weight:25.7 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:This antibody is raised against a recombinant protein of DsRed-Express2 expressed in HEK293 cells.
- Purification:affinity chromatography
- Pk:1 ml
Type: Tag
Antigen: DsRed-Express2
Clonality: Monoclonal
Clone: OTI1C4 (formerly 1C4)
Conjugation: Magnetic beads
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: