Anti-AP3D1 Goat Polyclonal Antibody
Catalog # ORIGTA319417
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:adaptor-related protein complex 3, delta 1 subunit
- Antigen symbol:AP3D1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Reactivity:Chicken,Chimpanzee,Human
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:This affinity purified antibody was prepared from whole goat serum produced by repeated immunizations with a synthetic peptide corresponding aa 686-698 of Human AP3D1.
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: ELISA: 1:5000-1:20000; Western Blot: 1:500-1:2000; Immunoprecipitation: 1:100
Type: Primary
Antigen: AP3D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Chicken, Chimpanzee, Human