Anti-LRRTM3 Rabbit Polyclonal Antibody
Catalog # ORIGTA320177
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen symbol:LRRTM3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Form:Liquid
- Storage buffer:LRRTM3 Antibody is supplied in PBS containing 0,02% sodium azide.
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:LRRTM3 antibody was raised against a 18 amino acid synthetic peptide near the carboxy terminus of human LRRTM3.
- Purification:purified by affinity chromatography via peptide column
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 0.5 ?g/ml; Immunochemistry: 2.5 ?g/ml; Immunofluorescence: 20 ?g/ml
Type: Primary
Antigen: LRRTM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat