Order Entry
Italy
ContactUsLinkComponent
Anti-TAG 72-4 / CA 72-4 Mouse Monoclonal Antibody [clone: B72.3]
Anti-TAG 72-4 / CA 72-4 Mouse Monoclonal Antibody [clone: B72.3]
Catalog # ORIGTA190272
Supplier:  OriGene
Anti-TAG 72-4 / CA 72-4 Mouse Monoclonal Antibody [clone: B72.3]
Catalog # ORIGTA190272
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    TAG 72-4
  • Clonality:
    Monoclonal
  • Clone:
    B72.3
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Antigen synonyms:
    CA 72-4
  • Storage buffer:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    –20 °C
  • Immunogen:
    Membrane-enriched fraction of a human breast carcinoma liver metastasis
  • Purification:
    purified by affinity chromatography
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Recommended Dilutions: Immunochemistry: 1:150

Type: Primary
Antigen: TAG 72-4 / CA 72-4
Clonality: Monoclonal
Clone: B72.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human