Anti-RASGRP3 Mouse Monoclonal Antibody [clone: 2G7]
Catalog # ORIGCF801465
Nome del fornitore: OriGene
Specifiche
- Antibody type:Primary
- Antigen name:RAS guanyl releasing protein 3 (calcium and DAG-regulated)
- Antigen symbol:RASGRP3
- Clonality:Monoclonal
- Clone:2G7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Amino acid number:380 to 690
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:78.2 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Human recombinant protein fragment corresponding to amino acids 380-690 of human RASGRP3 (NP_733772) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifiche
Informazioni su questo articolo
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150
Type: Primary
Antigen: RASGRP3
Clonality: Monoclonal
Clone: 2G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human