Rapid endotoxin detection, Pyrosate® kit
Certificates
About this item
The Pyrosate kit is a rapid gel-clot test that contains a endotoxin tube (PPC) matched to the sample (SPL) tube for each sensitivity. This feature is unique to the Pyrosate assay and ensures the validity of the test. The endotoxin tube (PPC) assures that the sample does not interfere with the test, ruling out false negatives. Pyrosate is formulated to eliminate false positives due to ß-D-glucans. This endotoxin specific reagent does not require additional blocking buffers.
- The test may be as short as 30 minutes, depending on the sensitivity
Frequently Bought Together