Culture media in bottles
Supplier: Dr. Möller & Schmelz
Certificates
About this item
These sterile ready-to-use media are filled in autoclavable and break-proof polycarbonate bottles (50 and 250 ml). The composition of the various types of media, comply with the formulations specified in the pertinent standards and regulations. Whereas the liquid media can be used immediately, the agar media first have to be remelted in a water bath at 90 to 100 °C. Once they have cooled down to around 50 °C you can pour agar plates in different sizes and thicknesses to meet your individual demands.
- 4×250 ml in polycarbonate bottles
Frequently Bought Together