Order Entry
Italy
ContactUsLinkComponent
Blades for safety knives, No. 45
Blades for safety knives, No. 45
Catalog # 233-2211
Supplier:  MARTOR ARGENTAX
undefined
Blades for safety knives, No. 45
Catalog # 233-2211
Supplier:  MARTOR ARGENTAX
Specifications
  • Description:
    Industrial blade no. 45, 4-times usable blade, 2-sided ground edge
  • L×W:
    39,0×18,4 mm
  • Material:
    Steel
  • Thickness:
    0,3 mm
  • Pk:
    10
Specifications
About this item
Industrial blade for use with safety knives and cutters.
  • Two-sided ground-edge
  • Single bevel edge-ground
  • Four-times usable blade
Mountable in:
ARGENTAX CLEVERCUT
SECUMAX EASYSAFE
SECUPRO OPTISAFE
SECUNORM HANDY
SECUNORM HANDYCUT
SECUNORM 175