Order Entry
Italy
ContactUsLinkComponent
Water purification system, Water Pro RO
Water purification system, Water Pro RO
Catalog # LBCP9075030
Supplier:  LABCONCO
undefined
Water purification system, Water Pro RO
Catalog # LBCP9075030
Supplier:  LABCONCO

Specifications

  • Flow rate:
    1,1 l/min
  • Description:
    WaterPro RO System
  • Model:
    WaterPro
  • Feed water:
    Potable (tap)
  • Weight:
    40,8 kg
  • W×D×H:
    790×193×782 mm
  • Pk:
    1

Specifications

About this item

WaterPro RO stations feature large capacity filters and membrane to deliver high-quality reverse osmosis (RO) purified water (type III). RO purified water is ideal for laboratory applications such as reagent preparation and glassware rinsing. The units be connected to glassware washers to provide purified rinse water.

  • Timed dispense from 1 to 99,9 minutes for unattended operation
  • Attractive and durable Type 304 stainless steel front panel; epoxy-coated steel cabinet
  • Large capacity white polypropylene bowls with threaded heads
  • Integral 17 liter PE storage tank
  • Dispensing valve

Water may be dispensed manually from a valve or optional dispensing gun. Water from the valve may also be activated automatically by using the exclusive timed dispense feature that allows for unattended operation.