Anti-IgG Goat Polyclonal Antibody (Europium 1024)
Supplier: Columbia Biosciences
Certificates
About this item
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: Europium 1024
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- COBSD17-112-1MG
- COBSD17-112-50
- COBSD17-112-20
- Antigen symbol:
- IgG
- IgG
- IgG
- Antibody type:
- Secondary
- Secondary
- Secondary
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Conjugation:
- Europium 1024
- Europium 1024
- Europium 1024
- Reactivity:
- Mouse
- Mouse
- Mouse
- Host:
- Goat
- Goat
- Goat
- Isotype:
- IgG
- IgG
- IgG
- Cross adsorption:
- Bovine, Rabbit
- Bovine, Rabbit
- Bovine, Rabbit
- Storage temperature:
- Cool
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
- Ice
Specifications
Frequently Bought Together