Order Entry
Italy
ContactUsLinkComponent
 
Anti-c-Myc tag Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 9E10]
  COBSAP-1714
Anti-c-Myc tag Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 9E10]
  COBSAP-1714

 

  • Antibody type:
    Tag
  • Antigen name:
    v-myc avian myelocytomatosis viral oncogene homolog
  • Antigen symbol:
    MYC
  • Clonality:
    Monoclonal
  • Clone:
    9E10
  • Conjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    none
  • Gene ID:
    4609
  • Antigen synonyms:
    MRTL|bHLHe39|c-Myc|MYCC
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Tag
Antigen: c-Myc tag (c-Myc tag)
Clonality: monoclonal
Clone: 90000000000
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: