Order Entry
Italy
ContactUsLinkComponent
Anti-IgG F(c) Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Anti-IgG F(c) Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # IMMRIR1739
Supplier:  ImmunoReagents
Anti-IgG F(c) Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # IMMRIR1739
Supplier:  ImmunoReagents

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG F(c)
  • Clonality:
    Polyclonal
  • Conjugation:
    TRITC (Tetramethylrhodamine Isothiocyanate)
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    F(c)
  • Cross adsorption:
    Yes
  • Format:
    Purified IgG
  • Form:
    Lyophilized
  • Antibody fragment:
    F(c)
  • Storage buffer:
    10 mM Sodium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 1 % (w/v) BSA, Protease/IgG free
  • Storage temperature:
    2…8 °C
  • Concentration:
    1,0 mg/ml (E 1% at 280 nm = 13,0)
  • Shipping temperature:
    2…8 °C
  • Immunogen:
    Human IgG, Fc fragment
  • Tested applications:
    Immunomicroscopy
  • Purification:
    Affinity purified using solid phaseHuman IgG
  • Size:
    1,0 mg
  • Pk:
    1 mg

Specifications

About this item

TRITC (Tetramethylrhodamine isothiocyanate) is a popular rhodamine derivative pink in color but emits a red-orange color upon Emission at 580nm. TRITC is frequently paired with FITC in double labeling experiments and conjugated antibodies are often used in immunofluorescence microscopy, flow cytometry, FLISA (fluorescent ELISA) and high throughput screening assays. These conjugates are best excited with the 532 nm spectral line of the He-Ne laser and are also recommended as a second color detection reagent in in situ hybridization applications. (Excitation/Emission = 555 nm / 580 nm Emission Color = Orange to Red (Similar Dyes: Alexa Fluor 555, Cy3, DyLight 550).

  • Origin: US

ImmunoReagents antibodies are suitable for most immunoassay formats and purified, cross adsorbed and conjugated based on the final product specfications.

Products will be accompanied with a standard product CoA to include application, purity and cross reactivity profiles.

Certifications: (EC) 142/2011 approved by APHIS, Reference Number is NC-TEC-0002.
Ordering information: Products can by customized based on specific assay requirements: contact your VWR representative for more information.

Type: Secondary
Antigen: IgG F(c)
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human