Anti-TBL2 Rabbit Polyclonal Antibody
Catalog # USBIW1002-70B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:transducin (beta)-like 2
- Antigen symbol:TBL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- ChIP:Yes
- Form:liquid
- Amino acid number:800 to 852
- Storage buffer:Tris-citrate/phosphate, pH 7,5, 0,01% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Tested applications:Chromatin
- Purification:purified by immunoaffinity chromatography
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Immunofluorescence: 1:100-1:500; Western Blot: 1:1000
Type: Primary
Antigen: TBL2 (transducin (beta)-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat