Anti-Transcobalamin 1 Chicken Polyclonal Antibody
USBIT8193
- Antibody type:Primary
- Antigen symbol:Transcobalamin 1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- Isotype:IgY
- Reactivity:Human
- Western blot:Yes
- Form:liquid
- Amino acid number:191 to 433
- Storage buffer:PBS, pH 7,2, 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:50 µg
- Pk:50 µG
Recommended dilution: Western Blot: 1:500
Type: Primary
Antigen: Transcobalamin 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human