Order Entry
Italy
ContactUsLinkComponent
Anti-TIP 39 Rabbit Polyclonal Antibody
Anti-TIP 39 Rabbit Polyclonal Antibody
Catalog # USBIT5499-87B-100
Supplier:  US Biological
Anti-TIP 39 Rabbit Polyclonal Antibody
Catalog # USBIT5499-87B-100
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    TIP 39
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Bovine,
    Dog
  • Format:
    Serum
  • Form:
    lyophilized powder
  • Storage buffer:
    PBSpH 7,2.
  • Sequence:
    SLALADDAAFRERARLLAALERRHWLNSYMHKLLVLDAP
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Tested applications:
    Radio Immunoassay
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Recommended dilution: ELISA: 1:4000; Radio Immunoassay: 1:2000

Type: Primary
Antigen: TIP 39
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Dog, Human