Anti-TCR, V beta 3 Mouse Monoclonal Antibody (Biotin) [clone: 5C95]
USBIT2040-17B
- Antibody type:Primary
- Antigen symbol:TCR V beta 3
- Clonality:Monoclonal
- Clone:5C95
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:liquid
- Storage buffer:PBS, pH 7,5, 0,2% BSA, 0,04% sodium azide,5% glycerol.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:120 Tests
- Pk:120 Tests
Recommended dilution: Flow Cytometry: 1:50
Type: Primary
Antigen: TCR, V beta 3
Clonality: monoclonal
Clone: 5C95
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human