Anti-TCR, alpha, beta, V beta 2 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 5K83]
Catalog # USBIT2040-02W3
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:TCR alpha/beta/V beta 2
- Clonality:Monoclonal
- Clone:5K83
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Chicken
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide and a stabilizing agent.
- Storage temperature:Cool
- Concentration:~0,1 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: TCR, alpha, beta, V beta 2
Clonality: monoclonal
Clone: 5K83
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Chicken