Anti-TCR, alpha, beta, V beta 1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 5K82]
USBIT2040-02V1
- Antibody type:Primary
- Antigen symbol:TCR alpha/beta/V beta 1
- Clonality:Monoclonal
- Clone:5K82
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Chicken
- Form:liquid
- Storage buffer:PBS, 0,1% sodium azide
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified
- Size:500 µg
- Pk:500 µG
Type: Primary
Antigen: TCR, alpha, beta, V beta 1
Clonality: monoclonal
Clone: 5K82
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Chicken