Anti-SYP Mouse Monoclonal Antibody [clone: 27G12]
USBIS9500-40D
- Antibody type:Primary
- Antigen name:Synaptophysin
- Antigen symbol:SYP
- Clonality:Monoclonal
- Clone:27G12
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Format:Supernatant
- Form:liquid
- Storage buffer:15mMsodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:1 ml
- Pk:1 mL
Recommended dilution: Western Blot: 1:500; Immunochemistry: 1:100-1:200
Type: Primary
Antigen: SYP (synaptophysin)
Clonality: Monoclonal
Clone: 27G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human