Anti-SYK Mouse Monoclonal Antibody [clone: 11D111]
Catalog # USBIS9100-01X
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Spleen Tyrosine Kinase
- Antigen symbol:SYK
- Clonality:Monoclonal
- Clone:11D111
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized powder
- Modification:phospho-Tyr525, phospho-Tyr526
- Storage buffer:PBS, pH 7,4, 5% trehalose.
- Storage temperature:Cool
- Concentration:~0,5 mg/ml (after reconstitution)
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: Western Blot: 0.1 ?g/ml
Type: Primary
Antigen: SYK (spleen tyrosine kinase)
Clonality: Monoclonal
Clone: 11D111
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human